Learn More
Triethyl borate, 97%, Thermo Scientific Chemicals
£72.23 - £260.30
Chemical Identifiers
CAS | 150-46-9 |
---|---|
Molecular Formula | C6H15BO3 |
Molecular Weight (g/mol) | 145.99 |
MDL Number | MFCD00009073 |
InChI Key | AJSTXXYNEIHPMD-UHFFFAOYSA-N |
Synonym | triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl esterShow More |
PubChem CID | 9009 |
ChEBI | CHEBI:38916 |
IUPAC Name | triethyl borate |
SMILES | B(OCC)(OCC)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031490
|
Thermo Scientific Acros
139582500 |
250 mL | Glass bottle |
£72.23
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626714
|
Thermo Scientific Acros
139580010 |
1 L | Glass bottle |
£260.30
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
150-46-9 | |
145.99 | |
AJSTXXYNEIHPMD-UHFFFAOYSA-N | |
9009 | |
triethyl borate |
C6H15BO3 | |
MFCD00009073 | |
triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl esterShow More | |
CHEBI:38916 | |
B(OCC)(OCC)OCC |
Specifications
150-46-9 | |
Colorless | |
117.0°C to 118.0°C | |
Authentic | |
C6H15BO3 | |
1.3720 to 1.3740 | |
01, 335 | |
triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester | |
AJSTXXYNEIHPMD-UHFFFAOYSA-N | |
triethyl borate | |
9009 | |
145.99 | |
Liquid |
-84.5°C | |
0.8580g/mL | |
11°C | |
96% min. (GC) | |
(C2H5O)3B | |
MFCD00009073 | |
0.858 | |
Solubility in water: decomposes | |
B(OCC)(OCC)OCC | |
145.99 | |
CHEBI:38916 | |
97% | |
Triethyl borate, 97% |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-760-9
missing translation for 'rtecsNumber' : ED5075000
missing translation for 'tsca' : TSCA
- Triethyl borate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.