missing translation for 'onlineSavingsMsg'
Learn More
Triethylene glycol, 99%, Thermo Scientific Chemicals Product Code.: 10041490

Triethylene glycol, 99%, Thermo Scientific Chemicals

Product Code. 10041490
2.5 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10041490

Brand: Thermo Scientific Acros 139590025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
MDL Number MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
Synonym triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanolShow MoreShow Less
PubChem CID 8172
ChEBI CHEBI:44926
SMILES OCCOCCOCCO

Specifications

Melting Point -7°C
Color Yellow
Density 1.1200g/mL
Boiling Point 285°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula HOCH2CH2OCH2CH2OCH2CH2OH
Refractive Index 1.4545 to 1.4565
Quantity 2.5 L
Beilstein 01, IV, 2400
Merck Index 15, 9835
Specific Gravity 1.12
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,benzene and toluene,sparingly soluble in ether,insoluble in petroleum ether,aliphatic,hydrocarbons
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethanol
Viscosity 48 mPa.s (20°C)
Formula Weight 150.17
Vapor Pressure <0.01mbar at 20°C
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material Triethylene glycol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463442 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number139590010,139590025,139590100
Lot NumberA0424493 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number139590010,139590025,139590100
Lot NumberA0456095 Certificate TypeCertificate of Analysis Date07/09/2023 Catalog Number139590010,139590025,139590100
Lot NumberA0451884 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number139590010,139590025,139590100
Lot NumberA0401273 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number139590010,139590025,139590100
Product Identifier
  • Triethylene glycol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethylene glycol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.