missing translation for 'onlineSavingsMsg'
Learn More
Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic acid, 98%, Thermo Scientific Chemicals Product Code.: 11419980

Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic acid, 98%, Thermo Scientific Chemicals

Product Code. 11419980
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11419980

Brand: Thermo Scientific Alfa Aesar 044737.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869-52-3
Molecular Formula C18H30N4O12
Molecular Weight (g/mol) 494.454
MDL Number MFCD00004290
InChI Key RAEOEMDZDMCHJA-UHFFFAOYSA-N
Synonym ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acidShow MoreShow Less
PubChem CID 70088
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Quantity 10 g
Formula Weight 494.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS10K011 Certificate TypeCertificate of Analysis Date20/06/2024 Catalog Number044737.09,044737.04
Lot NumberS03K032 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number044737.09,044737.04
Lot NumberN25F038 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044737.09,044737.04
Lot NumberN21G034 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044737.09,044737.04
Lot NumberL15Q061 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044737.09,044737.04
compliance-icons
Product Identifier
  • Triethylenetetramine hexaacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Triethylenetetraminehexaacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.