missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, For protein sequencing, Honeywell Fluka™

Purified by redistillation, for protein sequencing,≥99%

£56.10 - £401.08

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15632970
View Documents Promotion Details
Honeywell-Fluka
299537-25G
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15652970
View Documents Promotion Details
Honeywell-Fluka
299537-50G
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15622970
View Documents Promotion Details
Honeywell-Fluka
299537-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15642970
View Documents Promotion Details
Honeywell-Fluka
299537-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

76-05-1
114.02
DTQVDTLACAAQTR-UHFFFAOYSA-N
6422
trifluoroacetic acid
C2HF3O2
MFCD00004169
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
CHEBI:45892
OC(=O)C(F)(F)F

Specifications

76-05-1
CF3COOH
2699
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
OC(=O)C(F)(F)F
114.02
CHEBI:45892
≥99%
>100°C (>212°F)
97.5mmHg (20°C)
C2HF3O2
MFCD00004169
742035
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid
6422
114.02g/mol
1.489g/mL at 20°C
3.9 (vs air)
Trifluoroacetic Acid
Videos
Safety and Handling

Safety and Handling

P273-P280-P305 + P351 + P338-P310

H314-H332-H412

missing translation for 'einecsNumber' : 200-929-3

missing translation for 'rtecsNumber' : AJ9625000

SDS
Documents

Documents

Product Certifications