missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, HPLC Grade, J.T. Baker™
High purity solvents for use in analytical and preparative separations
£687.00 - £3715.00
Chemical Identifiers
CAS | 76-05-1 |
---|---|
Molecular Formula | C2HF3O2 |
Molecular Weight (g/mol) | 114.02 |
MDL Number | MFCD00004169 |
InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
PubChem CID | 6422 |
ChEBI | CHEBI:45892 |
IUPAC Name | trifluoroacetic acid |
SMILES | OC(=O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15538624
|
Avantor J.T.Baker
9470-01 |
70 mL | Can | N/A | |||||
15548624
|
Avantor J.T.Baker
9470-02 |
900 mL | Narrow Mouth Amber Glass Bottle |
£3715.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
trifluoroacetic acid |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 | |
OC(=O)C(F)(F)F |
Specifications
76-05-1 | |
MFCD00004169 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
trifluoroacetic acid | |
6422 | |
114.03 | |
1L = 1.53kg |
C2HF3O2 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
HPLC | |
Trifluoroacetic Acid, Gradient Tested for HPLC Peptide and Protein Analysis |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- trifluoroacetic acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title