Learn More
Triisopropylphosphine, 90+%, Thermo Scientific Chemicals
£30.30 - £308.00
Chemical Identifiers
CAS | 6476-36-4 |
---|---|
Molecular Formula | C9H21P |
Molecular Weight (g/mol) | 160.241 |
MDL Number | MFCD00015029 |
InChI Key | IGNTWNVBGLNYDV-UHFFFAOYSA-N |
Synonym | triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458 |
PubChem CID | 80969 |
IUPAC Name | tri(propan-2-yl)phosphane |
SMILES | CC(C)P(C(C)C)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15411337
|
Thermo Scientific Alfa Aesar
033572.02 |
0.5 g |
£30.30
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421337
|
Thermo Scientific Alfa Aesar
033572.04 |
2 g | N/A | ||||||
15431337
|
Thermo Scientific Alfa Aesar
033572.09 |
10 g |
£308.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triisopropylphosphine is used as a ligand in organometallic chemistry. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals. It is also used to produce Chlor(triisopropyl)phosphonium-dimesylamid at temperature of 40°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6476-36-4 | |
160.241 | |
IGNTWNVBGLNYDV-UHFFFAOYSA-N | |
80969 | |
CC(C)P(C(C)C)C(C)C |
C9H21P | |
MFCD00015029 | |
triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458 | |
tri(propan-2-yl)phosphane |
Specifications
6476-36-4 | |
176°C to 178°C | |
Pungent | |
1.467 | |
UN2845 | |
triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458 | |
IGNTWNVBGLNYDV-UHFFFAOYSA-N | |
tri(propan-2-yl)phosphane | |
80969 | |
≥90% | |
Triisopropylphosphine |
0.839 g/mL | |
-17°C (1°F) | |
C9H21P | |
MFCD00015029 | |
Air Sensitive | |
Soluble in alkanes. | |
CC(C)P(C(C)C)C(C)C | |
160.241 | |
160.24 | |
Liquid |
Safety and Handling
GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P222-P260-P264b-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H250-H314
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 229-333-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only