Learn More
Thermo Scientific Chemicals Trimethoprim, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced • Trimethoprim has antibacterial and antiprotozoal properties
Applications
• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects
• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression

Specifications
Specifications
Melting Point | 234.0°C to 236.0°C |
Loss on Drying | 0.5% max. (105°C, 4 h) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 5 g |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 290.32 |
Percent Purity | 98% |
Chemical Name or Material | Trimethoprim |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468525 | Certificate of Analysis | 25/01/2025 | 455121000,455120250,455120050 |
A0465677 | Certificate of Analysis | 26/08/2024 | 455121000,455120250,455120050 |
A0429642 | Certificate of Analysis | 06/11/2023 | 455121000,455120250,455120050 |
A0421065 | Certificate of Analysis | 10/04/2023 | 455121000,455120250,455120050 |
A0448660 | Certificate of Analysis | 05/01/2023 | 455121000,455120250,455120050 |
Safety and Handling
- Trimethoprim
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.