missing translation for 'onlineSavingsMsg'
Learn More
Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals Product Code.: 10511771

Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 10511771
250 g, Glass bottle
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10511771

Brand: Thermo Scientific Acros 140052500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3282-30-2
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000709
InChI Key JVSFQJZRHXAUGT-UHFFFAOYSA-N
Synonym pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchlorideShow MoreShow Less
PubChem CID 62493
IUPAC Name 2,2-dimethylpropanoyl chloride
SMILES CC(C)(C)C(Cl)=O

Specifications

Melting Point -56.0°C
Color Colorless to Pink
Density 0.9800g/mL
Boiling Point 105.0°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCOCl
Refractive Index 1.4110 to 1.4130
Quantity 250 g
Beilstein 02, 320
Fieser 01,1229; 08,404; 11,489
Specific Gravity 0.98
Solubility Information Solubility in water: hydrolysis., miscible with most common aprotic organic solvents
Viscosity 0.86 mPa.s (20°C)
Formula Weight 120.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethylacetyl chloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0445409 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number140050050,140052500,140050010,140051000
Lot NumberA0436965 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number140050050,140052500,140050010,140051000
Lot NumberA0445616 Certificate TypeCertificate of Analysis Date05/09/2022 Catalog Number140050050,140052500,140050010,140051000
Lot NumberA0444664 Certificate TypeCertificate of Analysis Date01/08/2022 Catalog Number140050050,140052500,140050010,140051000
Lot NumberA0422560 Certificate TypeCertificate of Analysis Date14/03/2022 Catalog Number140050050,140052500,140050010,140051000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.