missing translation for 'onlineSavingsMsg'
Learn More
Trimethyloxonium tetrafluoroborate, 96%, Thermo Scientific Chemicals Product Code.: 11412178

Trimethyloxonium tetrafluoroborate, 96%, Thermo Scientific Chemicals

Product Code. 11412178
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11412178

Brand: Thermo Scientific Alfa Aesar A15175.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.

Solubility
Soluble in nitrobenzene, nitromethane, chloroform, hot acetone, liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents.

Notes
Hygroscopic. Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420-37-1
Molecular Formula C3H9BF4O
Molecular Weight (g/mol) 147.91
MDL Number MFCD00011798
InChI Key CZVZBKHWOFJNCR-UHFFFAOYSA-N
Synonym trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborateShow MoreShow Less
PubChem CID 2735153
SMILES C[O+](C)C.F[B-](F)(F)F

Specifications

Melting Point ∽190°C (decomposition)
Assay Percent Range 96%
Quantity 10 g
Beilstein 3597303
Sensitivity Moisture sensitive
Solubility Information Soluble in nitrobenzene,nitromethane,chloroform,hot acetone,liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents.
IUPAC Name trimethyloxidanium;tetrafluoroborate
Formula Weight 147.91
Chemical Name or Material Trimethyloxonium Tetrafluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238225 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15175.30,A15175.18,A15175.09
Lot Number10218709 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15175.30,A15175.18,A15175.09
Lot NumberI6970A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15175.30,A15175.18,A15175.09
Lot Number10165010 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15175.30,A15175.18,A15175.09
Lot Number10114110 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15175.30,A15175.18,A15175.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethyloxonium tetrafluoroborate, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.