missing translation for 'onlineSavingsMsg'
Learn More
(Triphenylphosphoranylidene)acetaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 10158043

(Triphenylphosphoranylidene)acetaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 10158043
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10158043

Brand: Thermo Scientific Acros 307490250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2136-75-6
Molecular Formula C20H17OP
Molecular Weight (g/mol) 304.33
MDL Number MFCD00006994
InChI Key CQCAYWAIRTVXIY-UHFFFAOYSA-N
Synonym 2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5lShow MoreShow Less
PubChem CID 75051
IUPAC Name 2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
SMILES C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 185.0°C to 188.0°C
Color Brown to Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCHO
Quantity 25 g
Formula Weight 304.33
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material (Triphenylphosphoranylidene)acetaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0432097 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog Number307491000,307490250,307490050
Lot NumberA0397303 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number307491000,307490250,307490050
Lot NumberA0401948 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number307491000,307490250,307490050
Lot NumberA0402145 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number307491000,307490250,307490050
Lot NumberA0392738 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number307491000,307490250,307490050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Triphenylphosphoranylidene)acetaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.