Learn More
Triphosgene, 98%, Thermo Scientific Chemicals
Description
Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.
Solubility
Soluble in ether, ethanol, tetrahydrofuran, benzene, cyclohexane, chloroform, hexane, carbon tetrachloride, dichloromethane and chlorobenzene. Insoluble in water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 77°C to 91°C |
Density | 1.6 |
Boiling Point | 203°C to 206°C |
Quantity | 5 g |
UN Number | UN2928 |
Beilstein | 1787583 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ether,ethanol,tetrahydrofuran,benzene,cyclohexane,chloroform,hexane,carbon tetrachloride,dichloromethane and chlorobenzene. Insoluble in water. |
Formula Weight | 296.75 |
Percent Purity | 98% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10251936 | Certificate of Analysis | 09/08/2024 | A14932.22,A14932.14,A14932.06 |
10226614 | Certificate of Analysis | 17/10/2023 | A14932.22,A14932.14,A14932.06 |
10243180 | Certificate of Analysis | 13/10/2022 | A14932.22,A14932.14,A14932.06 |
10212634 | Certificate of Analysis | 24/05/2022 | A14932.22,A14932.14,A14932.06 |
5004L30X | Certificate of Analysis | 22/11/2021 | A14932.22,A14932.14,A14932.06 |
Safety and Handling


- Triphosgene
- Danger
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Triphosgene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.