missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS (Base), BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£185.00 - £580.00
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Synonym | trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15538844
|
Avantor J.T.Baker
X171-05 |
100 g | Poly Bottle |
£185.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15548844
|
Avantor J.T.Baker
X171-07 |
500 g | Poly Bottle |
£317.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15528844
|
Avantor J.T.Baker
X171-03 |
2.5 kg | Poly Pail |
£580.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
77-86-1 | |
121.136 | |
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
77-86-1 | |
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
BAKER ANALYZED™ ACS Reagent |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 | |
TRIS (Base) |