missing translation for 'onlineSavingsMsg'
Learn More
TRIS (Base), BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ Product Code.: 15548844

TRIS (Base), BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15548844
500 g, Poly Bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
This item is not returnable. View return policy

Product Code. 15548844

Brand: Avantor J.T.Baker X17107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris baseShow MoreShow Less
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Packaging Poly Bottle
Quantity 500 g
Formula Weight 121.14
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material TRIS (Base)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TRIS (Base), BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.