missing translation for 'onlineSavingsMsg'
Learn More

Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™

DNase-, RNase- and Protease-Free.

£112.00 - £2415.00

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, TrometamolShow MoreShow Less
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10103203
SDS View Documents Promotion Details
Fisher BioReagents
BP152-500
500 g Poly Bottle
£112.00
500g
Please to purchase this item. Need a web account? Register with us today!
10376743
SDS View Documents Promotion Details
Fisher BioReagents
BP152-1
1 kg Poly Bottle
£169.00
1kg
Please to purchase this item. Need a web account? Register with us today!
10724344
SDS View Documents Promotion Details
Fisher BioReagents
BP152-5
5 kg Poly Pail
£627.00
5kg
Please to purchase this item. Need a web account? Register with us today!
10667243
SDS View Documents
Fisher BioReagents
BP152-10
10 kg Poly Pail
£1440.00
10kg
Please to purchase this item. Need a web account? Register with us today!
10336793
SDS View Documents
Fisher BioReagents
BP152-25
25 kg Fiber Drum
£2415.00
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNase-, RNase- and Protease-Free.

Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

Product Identifier
  • Tris(hydroxymethyl)aminomethane

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.