missing translation for 'onlineSavingsMsg'
Learn More
Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10785341

Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10785341
1 kg
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 10785341

Brand: Fisher Chemical T/3710/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number 4679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Powder or Solid
Quantity 1 kg
Packaging Plastic powder jar
Formula Weight 121.14g/mol
Product Identifier
  • Tris(hydroxymethyl)aminomethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.