missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ TrypLE™ Select Enzyme (10X), no phenol red

TrypLE™ Select Enzyme (10X), no phenol red
£153.00 - £2370.00
Specifications
Chelators | EDTA |
---|---|
Classification | Animal Origin-free |
Concentration | 10 X |
Form | Liquid |
Product Type | Cell Culture Dissociation Reagent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10272625
![]() |
Gibco™
A1217701 |
100 mL |
£153.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10698308
![]() |
Gibco™
A1217703 |
20 x 100mL |
£2370.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10698118
![]() |
Gibco™
A1217702 |
500 mL |
£608.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A549, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE™ formulations for a range of cell culture applications.Compared to trypsin and other dissociation reagents, Gibco™ TrypLE™ Select is:
• Gentle on cells—maintains cell health for reproducible results
• Room-temperature stable—convenient to use
• Easy to use—substitutes directly into existing protocols, with no inactivation required
• Animal origin free (AOF)—pure, consistent, and effective
This TrypLE™ Select is modified as follows:
With
• EDTA
Without
• Phenol Red
The formulation of TrypLE™ solutions is available, although we keep the enzyme concentration confidential.
Gentle on cells
TrypLE™ Select, like trypsin, cleaves peptide bonds on the C-terminal sides of lysine and arginine. However, TrypLE™ Select's exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.
Room-temperature stable
TrypLE™ Select is room-temperature stable and ready to use. TrypLE™ Select cell dissociation reagents remain stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.
Easy to use
TrypLE™ Select can be substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE™ Select, avoiding the need for trypsin inhibitors, such as FBS.
Animal Origin Free (AOF)
Unlike porcine trypsin and other enzymes, TrypLE™ Select is free of animal-derived components, and is formulated on dedicated animal origin-free equipment.
Product Intended Use
For Research Use or Further Manufacturing. Not for use in diagnostic procedures. Customers using Gibco™ TrypLE™ Select in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).
cGMP Manufacturing and Quality System
Gibco™ TrypLE™ Select is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Concentrated forms of TrypLE™ can be aseptically diluted to 1x using a balanced salt solution containing 1 mM EDTA.
Specifications
EDTA | |
10 X | |
Cell Culture Dissociation Reagent | |
Energy efficient, Sustainable packaging | |
270 - 320 mOsm/kg | |
Gibco™ | |
Room Temperature Stable |
Animal Origin-free | |
Liquid | |
No Phenol Red | |
No Calcium, No Magnesium, No Sodium Phosphate | |
6.9 to 7.4 | |
Cell Culture Grade | |
In vitro Bioassay |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ TrypLE™ Select Enzyme (10X), no phenol red