missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Trypsin Inhibitor, soybeans Product Code.: 15443659

Thermo Scientific Chemicals Trypsin Inhibitor, soybeans

Product Code. 15443659
1 g
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy

Product Code. 15443659

Brand: Thermo Scientific Chemicals J60982.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits the pancreatic enzyme trypsin

The soybean trypsin inhibitor has been tested for cell culture applications. It has been used in a study to assess the potential application in animal cell culture of an alkaline protease from a non-toxigenic mangrove isolate of Vibri sp. V26. It has also been used in a study to improve the detection of fungi in eosinophilic mucin. It is an inhibitor of trypsin factor Xa, plasmin and neutrophil elastase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Dissolve in 1X PBS at 1 to 10mg/ml for stock solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9035-81-8

Specifications

Quantity 1 g
Description Off-white to Pale Brown
Content And Storage 2°C to 8°C
For Use With (Application) Inhibits the pancreatic enzyme trypsin
Product Type Signal Transduction Reagent
Form Powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ26J011 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberJ60982.03,J60982.MC
Lot NumberU23J013 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberJ60982.03,J60982.MC
Lot NumberX26I025 Certificate TypeCertificate of Analysis Date07/11/2022 Catalog NumberJ60982.03,J60982.MC
Lot NumberS15I013 Certificate TypeCertificate of Analysis Date27/06/2022 Catalog NumberJ60982.03,J60982.MC
Lot NumberP09I022 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ60982.03,J60982.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Trypsin Inhibitor, soybeans >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.