missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Tube Cleaning Brush

The Tube Cleaning Brush is ideal for cleaning zone samplers.

Brand:  Buerkle™ 5304-0100

47.07 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10578411

  • £54.50 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from PVC and stainless steel
  • Available in a range of lengths
Specifications

Specifications

Cleaning Brush
Used for Cleaning
100 cm
25 mm
Zone Sampler
PVC, Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Tube Cleaning Brush > Diameter: 25mm; Length: 100cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.