Learn More
Masterflex™ Turbo Prop Open Channel Flowmeter
Brand:  Masterflex™ 32922-01
Product Code. 11700099
  • £1720.00 / Each
Explore more special offers
Specifications
Paddle Wheel Flowmeter
Accurately measure water velocity in open channels and partially filled pipes
70°C
0.1 ft./sec.
Internal lithium Battery
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Turbo Prop Open Channel Flowmeter > Flow Rate Range: 0.3 to 15 ft./sec