missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween™ 20 (Polysorbate), BAKER ANALYZED™, J.T. Baker™, Avantor™
Laboratory Reagent
Brand: JT Baker 7374.1000
211.14 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
9005-64-5 | |
1 L | |
MFCD00165986 | |
1227.53 |
1.1 g/cm³ (20°C) | |
C58H114O26 | |
Polyoxyethylene sorbitan monolaurate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tween™ 20 (Polysorbate), BAKER ANALYZED™, J.T. Baker™, Avantor™ > 1 L
Spot an opportunity for improvement?Share a Content Correction