missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek 500 Industry Coveralls
GREENER_CHOICE

Provides an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 500 Industry Coveralls with Collar are composed of flash spun high density polyethylene.

Brand:  DuPont™ D13395377

620.33 GBP valid until 2025-12-31
Use promo code "24380" to get your promotional price.


 View more versions of this product

Product Code. 11753073

  • £835.00 / Pack of 100
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With collar
  • Internal stitched seams
  • Zipper made of TYVEK™ with flap
  • Elastic cuffs, ankles and waist
  • Antistatic treatment
  • Category III, Type 5/6
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industry Model Coveralls
HDPE
Elastic
Elastic
Zipper with Storm Flap
Category III, Type 5 and 6
EN 1073-2, EN 1149-5
For Maintenance and Dismantling Jobs in the Nuclear Industry, Pharmaceutical Manufacturing, or in Research and Biosecurity Laboratories.
Large
White
Elastic
Collared
Stitched
Standard fold
Elastic wrist, Zippered front
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek 500 Industry Coveralls > Size: Large

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.