missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ Collared Laboratory Coats with Press Studs, No Pockets
Offers enhanced protection for body parts that are more exposed to hazardous substances. Lab coat is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
£60.20 - £283.00
Specifications
Product Type | Chemical Resistant Lab Coat |
---|---|
Gender | Unisex |
Material | HDPE |
Color | White |
Closure Type | Press Studs |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11733123
|
DuPont™
D13495953 |
2X-Large |
£283.00
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
12302269
|
DuPont™
D13398975 |
X-Large |
£60.20
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
12392259
|
DuPont™
D13398884 |
Large |
£60.20
Pack of 10 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
Description
- Silicon non-added.
- With 5 press stud closures.
- Elasticated cuffs (not tunnelled).
- Stitched internal seams.
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols.
- Excellent barrier against fine particles and fibers (down to 1 micron in size).
- Ultra-low-linting.
- Antistatically treated.
- Applications include:
- Pharmaceutical Handling
- Chemical Protection
- Lead and Asbestos Abatement/Remediation
- General Maintenance/Operations
- Spray Painting and General Clean-up
Specifications
Chemical Resistant Lab Coat | |
HDPE | |
Press Studs | |
Limited-use | |
Chemicals | |
None | |
EN 1149-5 |
Unisex | |
White | |
Elastic | |
Non-sterile | |
Below Hip-length | |
Stitched, Internal |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek™ Collared Laboratory Coats with Press Studs, No Pockets