Learn More
UDP-GlcNAc Disodium Salt, MedChemExpress
MedChemExpress UDP-GlcNAc Disodium Salt (UDP-α-D-N-Acetylglucosamine Disodium Salt) is a donor substrate of O-GlcNAc transferase (OGT).
Brand: Medchem Express HY-112174
This item is not returnable.
View return policy
Description
UDP-GlcNAc Disodium Salt (UDP-α-D-N-Acetylglucosamine Disodium Salt) is a donor substrate of O-GlcNAc transferase (OGT).Specifications
UDP-GlcNAc Disodium Salt | |
White | |
C17H25N3Na2O17P2 | |
UDP-α-D-N-Acetylglucosamine Disodium Salt | |
O[C@H]1[C@H](N2C(NC(C=C2)=O)=O)O[C@H](COP(OP(O[C@@H]3[C@@H]([C@H]([C@H](O)[C@@H](CO)O3)O)NC(C)=O)(O[Na])=O)(O[Na])=O)[C@H]1O | |
651.32 | |
Research | |
Metabolism-protein/nucleotide metabolism |
91183-98-1 | |
Research | |
5 mg | |
DMSO : 250 mg/mL (383.84 mM; Need ultrasonic) ∣H2O : 125 mg/mL (191.92 mM; Need ultrasonic) | |
651.32 | |
98.0% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
UDP-GlcNAc Disodium Salt, MedChemExpress