missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ultimus Bottle Top Dispenser

Combines unique functionality, user-friendliness and performance. Fisherbrand™ Ultimus Bottle Top Dispenser offers dilution, rinising, dual liquid handling and re-circulation in a single unit.

Brand:  Fisherbrand™ ULTI10

 View more versions of this product

Product Code. 15558901

  • £484.00 / Each
Estimated Shipment: 14-08-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • With dual inlet technology
  • Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
  • Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
  • Specially designed adaptor allows 360° rotation providing full flexibility
  • PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
  • Adaptors to fit most of the laboratory reagent bottles
  • Colied extension tube for refilling
  • Calibration tool provided for convenient and quick in-lab user re-calibration

Specifications:

  • Piston: PTFE and ETP
  • Valve housing: PFA
  • Re-Circulation valve housing: PFA
  • Valve assembly: Borosilicate glass ball and seat
  • Discharge assembly: PTFE
  • Delivery and Inlet tube: FEP
  • Compatibility: With all reagents except HF
Specifications

Specifications

0.6% (+/-0.060)
Autoclavable (121°C, 15psi for 10-15 min.)
Borosilicate Glass
0.2 mL
Bottletop Dispenser
ISO 8655 standards
1 to 10 mL
ULT-10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Ultimus Bottle Top Dispenser > Increment: 0.2mL; Volume: 1 - 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.