Learn More
Description
- Dispenser roll with indicator strips 5m long
- pH range from 1 to 14
- 4/pack
Specifications
1 to 14 | |
Color Table | |
5 m |
Dispenser Roll | |
4/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.