Learn More
Fisherbrand™ Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
Brand:  Fisherbrand™ 0911R00031
 View more versions of this product
Product Code. 11703564
  • £21.70 / Each
Explore more special offers
Description
Description
  • Filter paper soaked with one or several colored indicators of pH
  • Economic range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
Specifications
5 to 9
1 Roll
5 m
Roll
Grade 911
7 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Universal pH Indicator Paper > pH Range: 5 to 9; Roll; Dimensions (L x W): 5m x 7mm