missing translation for 'onlineSavingsMsg'
Learn More

Uranium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

£65.65 - £157.00

Specifications

Concentration 1g/L
Solution Contains HNO3

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12993534
View Documents
Fisher Chemical
S 064 100
100 mL
£65.65
100mL
Please to purchase this item. Need a web account? Register with us today!
12903544
View Documents
Fisher Chemical
S 064 250
250 mL
£98.95
250mL
Please to purchase this item. Need a web account? Register with us today!
12913544
View Documents
Fisher Chemical
S 064 500
500 mL
£157.00
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

1g/L
HNO3
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Single-element Standard Solution for ICP. Uranium (U) 1000mg/l in HNO3 2% (S 064)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.