missing translation for 'onlineSavingsMsg'
Learn More
Urea, 98+%, Thermo Scientific Chemicals Product Code.: 11469493

Urea, 98+%, Thermo Scientific Chemicals

Product Code. 11469493
500 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11469493

Brand: Thermo Scientific Alfa Aesar A12360.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

Solubility
Soluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.

Notes
Stable at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 130°C to 135°C
Density 1.335
pH 9.2 to 9.5
Odor Ammonia-like
Quantity 500 g
Beilstein 635724
Merck Index 14,9867
Solubility Information Soluble in water,alcohol,methanol,glycerol,benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.
Formula Weight 60.06
Percent Purity ≥98%
Chemical Name or Material Urea
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253459 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog NumberA12360.0C,A12360.36,A12360.0E
Lot NumberX15F033 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberA12360.0C,A12360.36,A12360.0E
Lot Number10251454 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12360.0C,A12360.36,A12360.0E
Lot Number10250667 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12360.0C,A12360.36,A12360.0E
Lot Number10249327 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog NumberA12360.0C,A12360.36,A12360.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Urea, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.