missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 1 Safety Shoe S1

Sporty, multifunctional safety shoe in loafer style

£113.00 - £118.00

Specifications

Certifications/Compliance ESD certified, EN ISO 20345: 2011 S1 SRC
Color Black
Design Loafer
Material Microvelour
Toe Material Composite
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15381133
View Documents
Uvex™
8543838
38
£114.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
15311143
View Documents
Uvex™
8543841
41
£118.00
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
15351143
View Documents
Uvex™
8543845
45
£113.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
Description

Description

  • Especially lightweight and flexible
  • 100% metal-free uvex xenova™ toe cap of the latest generation, compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Made with a new last technology for extraordinary wearing comfort
  • Optimized comfort thanks to uvex climazone technology
  • Breathable, perforated high tech micro suede upper
  • Optimum ventilation thanks to Distance Mesh lining material
  • Suitable for allergy sufferers as made of synthetic materials
  • Individually adjustable, elastic lace with quick release
  • Almost seamless shaft construction to avoid pressure points
  • Removable comfort footbed, antistatic, with moisture transport system and additional heel and forefoot cushioning
  • Soft padded collar and padded flap
  • Special double-density polyurethane sole with excellent cushioning properties and very good slip resistance, free from substances that interfere with the wetting properties of the paint such as silicones, plasticisers, etc.
  • Meets the ESD requirements: Bleeder resistance <35 megohms
  • Certified with uvex medicare for individual orthopedic insoles and individual outsole modifications such as outsole heights, inner or outer heel elevations, roll-off aids and other solutions
  • Item No. 8543.8
  • Width 11
  • Standard EN ISO 20345: 2011 S1 SRC
  • Specifications

    Specifications

    ESD certified, EN ISO 20345: 2011 S1 SRC
    Loafer
    Composite
    Unisex
    No
    Black
    Microvelour
    11 mm
    Trainers
    ESD Safety Shoe
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Uvex™ 1 Safety Shoe S1

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.