missing translation for 'onlineSavingsMsg'
Learn More
Uvex™ Single Use Overall Product Code.: 15303313
Add to basket

Uvex™ Single Use Overall

Product Code. 15303313 Shop All Uvex Products
Medium
Click to view available options
Size:
Medium
This item is not returnable. View return policy

Product Code. 15303313

Brand: Uvex™ 9871010

Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color White
For Use With (Application) Protection in the lab
Hood Type Standard
Material Microporous Polypropylene Laminate
Size Medium
Ankle Style Elastic
Disposable Single-use
Product Type Hooded coverall
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Uvex™ Single Use Overall >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.