Learn More
BD Vacutainer™ Serum Tubes
Used to obtain a serum sample. BD™ Vacutainer™ Serum Tubes are made of durable PET plastic with a Hemogard™ closure. It has silica particles added to trigger coagulation, and are marked CAT and CE.
Brand: BD VS368975
Description
In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer™ plastic serum tubes have silica particles added for this purpose.
- These tubes are marked with the acronym CAT (Clot Activator Tube)
- Recommended minimum time for coagulation from patients who have not been treated with anticoagulants is 60 minutes
- Centrifuging conditions: ≤1300 g for 10 minutes at 18–25°C
Specifications
Serum Tube | |
PET | |
4 mL | |
13 x 75 mm | |
Block |
Silica | |
Hemogard | |
Red | |
Used to obtain a serum sample | |
CE, CAT |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.