Learn More
BD Vacutainer™ Serum Tubes
Used to obtain a serum sample. BD™ Vacutainer™ Serum Tubes are made of durable PET plastic with a Hemogard™ closure. It has silica particles added to trigger coagulation, and are marked CAT and CE.
Brand: BD 367837
Includes: Pack of 100 tubes.
Description
In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer™ plastic serum tubes have silica particles added for this purpose.
- These tubes are marked with the acronym CAT (Clot Activator Tube)
- Recommended minimum time for coagulation from patients who have not been treated with anticoagulants is 60 minutes
- Centrifuging conditions: ≤1300 g for 10 minutes at 18–25°C

Specifications
Serum Tube | |
PET | |
6 mL | |
13 x 100 mm | |
Used to obtain a serum sample | |
CE, CAT |
Silica | |
Hemogard | |
Red | |
Pk. of 100 tubes | |
Block |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- TUBE PLN PLH 13X100 6.0 BLBLCE RD
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : quartz
Your input is important to us. Please complete this form to provide feedback related to the content on this product.