missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ Jeio Tech 665L Vacuum Oven (OV-12)

Optimally constructed vacuum oven for separating solvents from solutions or dry solvents with high boiling points. Medline Scientific™ Jeio Tech 665L Vacuum Oven (OV-12) is suitable for a variety of agricultural, plant-related, and drug metabolism applications.

Brand:  Medline Scientific™ AAH13125K

Product Code. 12949948

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Microprocessor PID control
  • Temperature calibration
  • Uniform heat distribution from block type heater attached to the chamber exterior to the anodized aluminum shelves
  • Intuitive control panel with bright LED display (1°C resolution).
  • Dual wait on/off timer modes (1 min. to 99 hr. 59 min.)
  • Automatic tuning
  • Convenient pre-setting function for three most commonly used temperature settings
  • Viton (fluorine rubber) door gasket also available for acidic applications (optional).
  • Heat-tempered glass window provides clear inside view of the chamber
  • Tight vacuum seal thanks to spring-loaded glass door, silicone gasket, and a push-button latch handle
  • Separate vacuum and vent ports
  • RS-232 interface for external control and data collection
  • Keypad lock function prevents accidental changes during operation
  • Automatic run after power interruption
  • Over-temperature and over-current protection
Specifications

Specifications

65 L
OV-12
405 x 402 x 402 mm
595 mm
557 mm
230 V 50/60 Hz
Ambient +5° to 250°C
3
557 x 780 x 595 mm
402 mm
405 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers