Learn More
Memmert™ Vacuum Oven Thermoshelf
Brand:  Memmert™ VO101B00735
 View more versions of this product
Product Code. 11911459
  • £694.00 / Each
Estimated Shipment: 13-08-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Calibration certificate
Shelf
Local Temperature Sensing (Pt100, 4-Wire-Circuit)
Model 500
Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control)
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Vacuum Oven Thermoshelf > Stainless Steel; For use with Model 500