missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

Product Code. 11384919
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment

Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Boiling Point 140°C (13mmHg)
Melting Point 256°C to 259°C
Physical Form Powder
Quantity 50 g
UN Number UN3285
Solubility Information Moderately soluble in acetone, ether and chloroform; Soluble in ethanol and benzene
Formula Weight 265.16
Density 1.4 g/mL
Chemical Name or Material Vanadium (IV) oxide bis (2,4-pentanedionate)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'