missing translation for 'onlineSavingsMsg'
Learn More
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals Product Code.: 10163880

Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals

Product Code. 10163880
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10163880

Brand: Thermo Scientific Acros 199590500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 256.0°C to 259.0°C
Color Blue-Green to Green
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Linear Formula (CH3COCH=C(CH3)O)2V=O
Quantity 50 g
Fieser 02,456; 03,331; 11,47; 13,54; 15,57; 15,186
Solubility Information Solubility in water: negligible. Other solubilities: soluble in methanol (6.4 g/100 mL), ethanol, ether, chloroform, acetone, dioxane, benzene (0.9 g/100 m, l), pyridine
Formula Weight 265.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Vanadyl(IV) acetylacetonate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0441772 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number199590500,199592500,199590010
Lot NumberA0451496 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number199590500,199592500,199590010
Lot NumberA0398975 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number199590500,199592500,199590010
Lot NumberA0395376 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number199590500,199592500,199590010
Lot NumberA0401641 Certificate TypeCertificate of Analysis Date20/10/2021 Catalog Number199590500,199592500,199590010
compliance-icons
Product Identifier
  • Vanadyl(IV) acetylacetonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.