Learn More
3M™ Versaflo™ Powered Air Respirator Starter Kit, TR-315E+
3M™ Versaflo Powered Air System TR-300+ combines performance, protection and functionality with modern, stylish, lightweight design and unrivalled comfort. It is the key to easy-to-use, versatile respiratory protection for particulate environments.
Brand: 3M™ 7100098908
759.95 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Simple starter kit with all the component parts to get you up and running. Simply add the headtop required
- Easy to use, versatile respiratory protection against dusts, mists, fumes and other particulate hazards
- Kit includes Turbo Unit TR-302E+, particulate filter, pre-filter, high-capacity battery, battery charger, length-adjusting breathing tube, air flow indicator, standard belt and spark arrestor
- Compatible headtop required
- Intelligent lithium ion battery quickly charges, has no memory effect and incorporates useful charge indicator
- Electronic audible and visual low air flow and low battery alarms as well as battery charge indicator /particulate filter loading status indicators and airflow indicator
- 3M™ Versaflo™ Powered Air Starter Kit TR-315+ contains TR-302E+ Turbo Unit, Particle Filter, Pre-Filter, Spark Arrestors, Standard Belt, High Capacity Battery, Battery Charger Kit and length adjusting Breathing Tube. Simply add your 3M™ Versaflo™ Headtop for complete respiratory protection.

Specifications
High Capacity Lithium Ion | |
190 L/min | |
TR-300 Turbo unit, P filter, pre-filter, spark arrestor, standard belt, high capacity battery, battery charger kit, and length adjusting breathing tube | |
Belt-mounted |
PAPR Assembly, No Headtop | |
Pharmaceutical manufacturing, woodworking, surface preparation, food and beverage, metal working, smelting | |
7100098908 | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
3M Powered and supplied air products
Your input is important to us. Please complete this form to provide feedback related to the content on this product.