Learn More
Invitrogen™ ViewRNA™ Temperature Validation Kit
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy.
Brand:  Invitrogen™ QV0523
Product Code. 16328755
  • £345.00 / Each
Explore more special offers
Description
Description
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy. This kit may be used for any ViewRNA Assay to evaluate the temperature of incubators or ThermoBrite systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Temperature Validation Kit
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number441815-000 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number440692-000 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number435988-000 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberQV0523
Lot Number412973-000 Certificate TypeCertificate of Analysis Date07/06/2024 Catalog NumberQV0523
Lot Number409764-000 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog NumberQV0523
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ ViewRNA™ Temperature Validation Kit > 1 Kit