missing translation for 'onlineSavingsMsg'
Learn More
Vinylboronic anhydride pyridine complex, 95%, Thermo Scientific Chemicals Product Code.: 11472110

Vinylboronic anhydride pyridine complex, 95%, Thermo Scientific Chemicals

Product Code. 11472110
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11472110

Brand: Thermo Scientific Alfa Aesar H28703.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent used for Suzuki-Miyaura cross-coupling, stereoselective synthesis via Palladium-catalyzed carboamination, Alkyl-connected 2-amino-6-vinylpurine (AVP) cross linking agent to cytosine base in RNA, Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media and Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for Suzuki-Miyaura cross-coupling, stereoselective synthesis via Palladium-catalyzed carboamination, Alkyl-connected 2-amino-6-vinylpurine (AVP) cross linking agent to cytosine base in RNA, Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media and Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers.

Solubility
Soluble in methanol.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95010-17-6
Molecular Formula C11H14B3NO3
Molecular Weight (g/mol) 240.667
MDL Number MFCD03839940
InChI Key YLHJACXHRQQNQR-UHFFFAOYSA-N
Synonym 2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1, trivinylboroxin pyridine complex, vinylboronic anhydride pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine, 2,4,6-trivinylcyclotriboroxane pyridine, vinylboronic anhydride pyridine, 2,4,6-trivinylcyclotriboroxane pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine complex, boroxin, triethenyl-, compd. with pyridine 1:1, pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinaneShow MoreShow Less
PubChem CID 2734806
IUPAC Name pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1

Specifications

Melting Point 44°C to 52°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol.
Formula Weight 240.67
Percent Purity 95%
Chemical Name or Material Vinylboronic anhydride pyridine complex

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238215 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH28703.06,H28703.03
Lot Number10182849 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28703.06,H28703.03
Lot Number10147472 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28703.06,H28703.03
Lot Number10219133 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28703.06,H28703.03
Lot Number10179790 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28703.06,H28703.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vinylboronic anhydride pyridine complex, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.