Learn More
Waters Corporation Viridis CSH FL-Phenyl Column
Designed and tested for SFC, the Viridis columns demonstrate a wide range of selectivity for greater resolution.
£964.50 - £25395.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
Endcapped | No |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
17113242
![]() |
Waters Corporation
186005688 |
50 mm | 3 mm | 5 μm |
£964.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17123242
![]() |
Waters Corporation
186005689 |
100 mm | 3 mm | 5 μm |
£1057.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17133242
![]() |
Waters Corporation
186005690 |
150 mm | 3 mm | 5 μm |
£1132.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17143242
![]() |
Waters Corporation
186005691 |
250 mm | 3 mm | 5 μm |
£1240.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17153242
![]() |
Waters Corporation
186005692 |
50 mm | 4.6 mm | 5 μm |
£982.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17163242
![]() |
Waters Corporation
186005693 |
100 mm | 4.6 mm | 5 μm |
£1074.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17173242
![]() |
Waters Corporation
186005694 |
150 mm | 4.6 mm | 5 μm |
£1143.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17183242
![]() |
Waters Corporation
186005695 |
250 mm | 4.6 mm | 5 μm |
£1252.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17193242
![]() |
Waters Corporation
186005700 |
50 mm | 19 mm | 5 μm |
£2700.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17103252
![]() |
Waters Corporation
186005701 |
100 mm | 19 mm | 5 μm |
£4263.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17113252
![]() |
Waters Corporation
186005702 |
150 mm | 19 mm | 5 μm |
£5355.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17123252
![]() |
Waters Corporation
186005703 |
250 mm | 19 mm | 5 μm |
£6021.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17133252
![]() |
Waters Corporation
186005704 |
50 mm | 30 mm | 5 μm |
£5814.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17143252
![]() |
Waters Corporation
186005705 |
75 mm | 30 mm | 5 μm |
£7215.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17153252
![]() |
Waters Corporation
186005706 |
100 mm | 30 mm | 5 μm |
£8686.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17163252
![]() |
Waters Corporation
186005707 |
150 mm | 30 mm | 5 μm |
£11835.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17173252
![]() |
Waters Corporation
186005708 |
250 mm | 30 mm | 5 μm |
£18615.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17183252
![]() |
Waters Corporation
186005709 |
50 mm | 50 mm | 5 μm |
£10548.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17193252
![]() |
Waters Corporation
186005710 |
100 mm | 50 mm | 5 μm |
£14478.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17103262
![]() |
Waters Corporation
186005711 |
150 mm | 50 mm | 5 μm |
£19650.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17113262
![]() |
Waters Corporation
186005712 |
250 mm | 50 mm | 5 μm |
£25395.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17103762
![]() |
Waters Corporation
186006543 |
150 mm | 2.1 mm | 5 μm |
£1132.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17113772
![]() |
Waters Corporation
186006567 |
50 mm | 2.1 mm | 1.7 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17123772
![]() |
Waters Corporation
186006568 |
75 mm | 2.1 mm | 1.7 μm |
£1183.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17133772
![]() |
Waters Corporation
186006569 |
100 mm | 2.1 mm | 1.7 μm |
£1240.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17143772
![]() |
Waters Corporation
186006570 |
150 mm | 2.1 mm | 1.7 μm |
£1309.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17153772
![]() |
Waters Corporation
186006571 |
50 mm | 3 mm | 1.7 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17163772
![]() |
Waters Corporation
186006572 |
75 mm | 3 mm | 1.7 μm |
£1183.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17173772
![]() |
Waters Corporation
186006573 |
100 mm | 3 mm | 1.7 μm |
£1240.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes – without the use of mobile-phase additives – even for well-retained basic achiral compounds.
Specifications
6000 psi (415 Bar) | |
130 Å | |
No |
Spherical | |
1/Ea. | |
Column |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.