missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Vivaspin™ 20 Sample Concentrators

Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 20 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.

Brand:  Cytiva 28-9323-60

161.50 GBP valid until 2025-12-31
Use promo code "24320" to get your promotional price.


 View more versions of this product

Product Code. 10015804

  • £170.00 / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.

  • Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
  • Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
  • Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
  • Compatible pH range: 1 to 9
  • Active membrane area: 6.0cm2

For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PES
6 cm2
5 to 20 mL
30 mm
10 kDa
25/Pk.
116 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Vivaspin™ 20 Sample Concentrators > Vivaspin 20, 10,000MW size

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.