missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin Filtrate
BIOPHARMA

The unique design of Vivaspin™ Filtrate ensures that the ultrafiltrate is collected above the membrane, ready for direct retrieval and analysis. Vivaspin filtrate is a ready-to-use centrifugal ultrafilter that is ideal for isolating small drug molecules, hormones and metabolites for analytical results you can rely on.

Brand:  Sartorius 13269---------GE

82.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 17918132

  • £89.20 / Pack of 12
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tame Tough Samples
  • More to Analyze
  • Interference-Free
  • Label Friendly
  • Direct Sample Retrieval
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Centrifuge Filter
Centrifuge
PES
2.5 mL
Solvent Absorption
Spin Column
Polystyrene, Polyethylene, SAN
100 kDa
Non-sterile
Filtrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Vivaspin Filtrate > Type:Centrifugal,Membrane Material:Polyethersulfone

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.