missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Selective allosteric potentiator of M4 acetylcholine receptors (EC50 values are 33 and 0.7 nM for ACh in the absence and presence of VU10010 respectively). Binds to an allosteric site on the receptor increasing affinity for ACh and coupling to G-proteins.

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 345.85 |
Percent Purity | >99% |
Chemical Name or Material | VU 10010 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VU 10010, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction