Learn More
Fisherbrand™ Wall Bracket for Additional Cartridge
For adding additional light water filtration and purification cartridges. Fisherbrand™ Wall Bracket for Additional Cartridge provides for safe and secure connection.
Brand: Fisherbrand™ SU
65.23 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
- Facilitates the water purification process
- Supplied in individual packs
Specifications
60 x 11 cm | |
Deionization Cartridges; Activated Carbon Cartridge; Micron Filter Cartridge; TOC Ion Exchange Cartridge; KCR Quick Coupling Kit | |
Wall Bracket |
Water purification | |
1 Each |