missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water Plate Count Prepared Agar, Bottled

Enumerate all culturable microorganisms from water using colony count technique with this nutritious and ISO compliant medium.

Brand:  Thermo Scientific™ BO0055R

96.44 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11958419

  • £110.24 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Enumerate all culturable microorganisms from water using colony count technique with Thermo Scientific™ Oxoid™ Water Plate Count Agar, ready prepared bottles. This nutritious, non-selective medium conforms to ISO 6222: 1999 (E) Water quality — Enumeration of culturable microorganisms — Colony count by inoculation in a nutrient agar culture medium1.

Waters of all kinds invariably contain a variety of microorganisms derived from various sources, such as soil and vegetation. Estimation of the overall numbers provide useful information for the assessment and surveillance of water quality. Separate counts are usually made of the microorganisms which are able to grow and form colonies on nutrient agar media at 36°C and 22°C. Colony counts are useful for assessing the integrity of ground water sources and the efficiency of water treatment processes such as coagulation, filtration and disinfection and provide an indication of the cleanliness and integrity of the distribution system. They can also be used to assess the suitability of a water supply for the preparation of food and drink to avoid contaminating the product with spoilage organisms. The main value of colony counts lies in the detection of changes from expected counts, based on frequent, long term monitoring. Any sudden increase in the count can be an early warning of serious pollution and a call for immediate investigation.1 Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Straw
Water Plate Count Agar (ISO)
United Kingdom
200.0g +2.5g
10/Pk.
2 to 25°C
250 mL sirop bottle
Agar
10 sirop bottles in a box
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4500546 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberBO0055R
Lot Number4500614 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberBO0055R
Lot Number4500547 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberBO0055R
Lot Number4498270 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberBO0055R
Lot Number4496462 Certificate TypeCertificate of Analysis Date09/03/2025 Catalog NumberBO0055R
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Water Plate Count Prepared Agar, Bottled >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.