missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Waterproof pH Pen

pH measurement is easier than ever with the Fisherbrand™ pH Pen. An economical range pocket-size tester designed for on-the-go pH measurements.

Brand:  Fisherbrand™ 16417270

65.87 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 16417270

  • £117.51 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Accurate and easy to use, these testers are ideal for quick pH measurements in hydroponic gardening, aquaculture, agriculture, pools, simple lab work and other water/wastewater applications.

  • ±0.1% accuracy across full pH range
  • Auto-buffer recognition and auto-calibration functions – quick, easy calibrations at the press of a button
  • Long-lasting sensor – large volume of polymer gel reference gives longer sensor lifespan, while single Kynar™ reference junction prevents clogging
  • Automatic temperature compensation and up to three calibration points for broadened accuracy throughout the pH range
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.1
0.0 to 14.0
LCD, 3.5 Digit with Annunciator
16.3 x 4.5 x 3 cm
Automatic Temperature Compensation (ATC)
Automatic
0.1
IP-67 Waterproof
90 g
4 x 1.5 V 'A76' Micro Alkaline Batteries
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Waterproof pH Pen > Dimensions: 16.3 x 4.5 x 3 cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.