Learn More
Testo™ Waterproof Surface Probe
Brand:  Testo™ 0609 1973
Product Code. 10478983
  • £234.00 / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Probe, Surface
EN 13485 and HACCP-compliant
40 sec.
-50°C to +400°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Testo™ Waterproof Surface Probe > Probe Diameter: 5mm