Learn More
Thermo Scientific™ WebSeal™ Sealing Tapes
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes.
Brand:  Thermo Scientific™ 60180-M143
166.90 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15132719
  • £197.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.
Easy, Consistent Sealing
  • Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Tapes seal onto the plate with a convenient, handheld applicator (available separately)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
100
Webseal
-70°C to +100°C
PES, Silicone
Non-sterile Sealing Tape
Pressure Sensitive
All Microplates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ WebSeal™ Sealing Tapes > PES/Silicone
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.