Learn More
Cytiva Whatman™ Gel Blotting Paper Grade GB003
A thick paper ideal for lysis/denaturation of colony or plaque lifts and Western blots
Brand: Cytiva 10427805
31.41 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
This item is not returnable.
View return policy
Description
- High absorbency
- Pure cellulose
- No contamination during transfers
- Fo chromatographic and blotting techniques
Specifications
150 mm | |
Suitable for (lysis/denaturation of colony or plaque lifts and Western blots) | |
GB003 |
100 mm | |
Cellulose |