Learn More
Cytiva Whatman™ Polycap™ HD 36 Disposable Capsule Filters
Ensures a finishing step yielding high purity final product. Whatman™ Polycap™ HD 36 Capsule Filters are suitable for filtering chemical solutions across wide temperature and pH ranges.
£54.80 - £289.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material (Membrane) | Monofilament Anisotropic Polypropylene |
For Use With (Application) | Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. |
Product Code | Brand | Pore Size | Inlet Style | Outlet Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Inlet Style | Outlet Style | Price | Quantity & Availability | |||
11314604
|
Cytiva
6703-3611 |
10 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
£82.95
Each |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11304604
|
Cytiva
6703-3610 |
1 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
£80.10
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11334604
|
Cytiva
6703-3650 |
5 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
£69.92
Each |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11324604
|
Cytiva
6703-3621 |
20 μm | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing | Stepped Barb for 6 to 10mm (1/4 to 3/8 in.) tubing |
£54.80
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11369114
|
Cytiva
2610T |
0.2 μm | FNPT | FNPT |
£289.00
Pack of 5 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11339124
|
Cytiva
2613T |
10 μm | FNPT | FNPT |
£274.00
Pack of 5 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11389114
|
Cytiva
2611T |
1 μm | FNPT | FNPT |
£274.00
Pack of 5 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11319124
|
Cytiva
2612T |
5 μm | FNPT | FNPT |
£289.00
Pack of 5 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11329124
|
Cytiva
2614T |
20 μm | FNPT | FNPT |
£265.00
Pack of 5 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11309124
|
Cytiva
2611 |
1 μm | Stepped Barb 10 to 12mm (3/8 to 1/2 in.) | Stepped Barb 10 to 12mm (3/8 to 1/2 in.) |
£245.00
Pack of 5 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Ensures a finishing step yielding high purity final product. Whatman™ Polycap™ HD 36 Capsule Filters are suitable for filtering chemical solutions across wide temperature and pH ranges.
Filtration of aqueous solutions, biologicals, inks and pigments, photographic emulsions, food and beverages, solvents. Offer broad capacity between gross paper filters and microporous membranes.- Integrity testable
- With inlet vent
- Filter Media: Monofilament anisotropic polypropylene (MAPP) filter
- Able to be sterilized by autoclaving with steam or ethylene oxide (EtO)
- Retention (with air purge), <2mL
- Max pressure: 4.1 bar (60 psi)
Specifications
Autoclavable | |
Monofilament Anisotropic Polypropylene | |
Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. |